siRNA Details
Gene Symbol: ZNF667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321356.1 | NP_001308285.1 | 3 | 659 | ||
XM_011527208.1 | XP_011525510.1 | 2 | 2303 | |||
GenBank mRNA | AK096513.1 | - | 3 | 588 | ||
AK126957.1 | - | 1 | 1298 | |||
AK127426.1 | - | 2 | 2303 |
