siRNA Details
Gene Symbol: PCGEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002769.1 | 3 | 974 | |||
GenBank mRNA | AF223389.1 | - | LIT1562 PCGEM1 |
3 | 974 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PCGEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002769.1 | 3 | 974 | |||
GenBank mRNA | AF223389.1 | - | LIT1562 PCGEM1 |
3 | 974 |