siRNA Details
Gene Symbol: LOC440896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015361.1 | 4 | 2888 | |||
GenBank mRNA | AK055091.1 | - | 4 | 2891 | ||
AK127288.1 | - | 5 | 4071 | |||
AY343901.1 | - | 3 | 292 | |||
AY343902.1 | - | 3 | 287 | |||
AY343903.1 | - | 3 | 292 | |||
BC032955.1 | - | 8 | 1921 | |||
BC036611.1 | - | 4 | 2577 |
