siRNA Details
Gene Symbol: LOC440896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015361.1 | 3 | 2461 | |||
GenBank mRNA | AK055091.1 | - | 3 | 2464 | ||
AK127288.1 | - | 3 | 3575 | |||
BC036611.1 | - | 2 | 2081 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC440896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015361.1 | 3 | 2461 | |||
GenBank mRNA | AK055091.1 | - | 3 | 2464 | ||
AK127288.1 | - | 3 | 3575 | |||
BC036611.1 | - | 2 | 2081 |