siRNA Details
Gene Symbol: FAR2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026758.2 | 9 | 2658 | |||
GenBank mRNA | AK125921.1 | - | 9 | 2660 | ||
BC132694.1 | - | 4 | 356 | |||
BC132696.1 | - | 4 | 356 | |||
BC144438.1 | - | 4 | 356 | 9052961 | ||
EU668357.1 | - | 1 | 545 |
