siRNA Details
Gene Symbol: ZNF619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026746.2 | 4 | 510 | |||
GenBank mRNA | AK304741.1 | - | 4 | 439 | ||
BC143331.1 | - | 2 | 191 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DC422729.1 | - | 4 | 439 | Chr. 3 - 40477184 - 40482340 [+] on Build GRCh38 |
