siRNA Details
Gene Symbol: PARVG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028907.1 | XP_016884396.1 | 5 | 774 | ||
XM_017028908.1 | XP_016884397.1 | 5 | 794 | |||
GenBank mRNA | AK293360.1 | - | 4 | 756 | ||
AK307294.1 | - | 5 | 973 | |||
AK307299.1 | - | 5 | 774 | |||
AK309348.1 | - | 4 | 771 | |||
AK310087.1 | - | 4 | 591 | |||
BX648942.1 | - | 1 | 1458 |
