siRNA Details
Gene Symbol: LOC400043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026656.1 | 2 | 747 | |||
GenBank mRNA | BC009385.2 | - | 2 | 720 | ||
BU508046.1 | - | 2 | 635 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC400043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026656.1 | 2 | 747 | |||
GenBank mRNA | BC009385.2 | - | 2 | 720 | ||
BU508046.1 | - | 2 | 635 |