siRNA Details
Gene Symbol: LINC01091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027105.2 | 3 | 223 | |||
NR_027106.1 | 2 | 136 | ||||
GenBank mRNA | BC033378.1 | - | 2 | 136 | ||
BC046914.1 | - | 2 | 136 | |||
BC053945.1 | - | 2 | 136 | |||
CK821624.1 | - | 3 | 227 |
