siRNA Details
Gene Symbol: RPL34-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026968.1 | 2 | 430 | |||
GenBank mRNA | AK094992.1 | - | 2 | 370 | ||
AK308624.1 | - | 3 | 496 | |||
BC118631.1 | - | 2 | 360 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RPL34-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026968.1 | 2 | 430 | |||
GenBank mRNA | AK094992.1 | - | 2 | 370 | ||
AK308624.1 | - | 3 | 496 | |||
BC118631.1 | - | 2 | 360 |