siRNA Details
Gene Symbol: MIR431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029965.1 | 1 | 51 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NR_029951.
