siRNA Details
Gene Symbol: ZNF519 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033354.1 | 6 | 1324 | |||
XM_017025562.1 | XP_016881051.1 | 6 | 10757 | |||
XM_017025563.1 | XP_016881052.1 | 7 | 6757 | |||
XM_017025564.1 | XP_016881053.1 | 6 | 6496 | |||
XM_017025565.1 | XP_016881054.1 | 7 | 6574 | |||
GenBank mRNA | AK096748.1 | - | 5 | 1161 | ||
BC037787.1 | - | 6 | 1292 |
