siRNA Details
Gene Symbol: DICER1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015415.1 | 3 | 360 | |||
GenBank mRNA | BC040596.1 | - | 1 | 1371 | ||
BI754942.1 | - | 3 | 286 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE788052.1 | - | 370 | Chr. - - [-] on Build |
