siRNA Details
Gene Symbol: DEFA10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029386.1 | 1, 2 | 176 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AY746435.1 | - | 1, 2 | 176 | Chr. 8 - 6968141 - 6969113 [-] on Build GRCh38 |
