siRNA Details
Gene Symbol: LINC00643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015358.2 | 2, 3 | 355 | |||
GenBank mRNA | AK311468.1 | - | 2, 3 | 559 | ||
BX248751.1 | - | 2, 3 | 233 |

More Information
Other Species Targeted

