siRNA Details
Gene Symbol: LINC00643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015358.2 | 4 | 588 | |||
GenBank mRNA | AK096461.1 | - | 1 | 110 | ||
AK311468.1 | - | 4 | 792 | |||
BX248751.1 | - | 4 | 466 |

More Information
Other Species Targeted

