siRNA Details
Gene Symbol: LINC01622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027115.3 | 4, 5 | 2020 | |||
NR_027116.2 | 3, 4 | 1913 | ||||
GenBank mRNA | BC037855.2 | - | 3, 4 | 1914 | ||
BC117448.1 | - | 4, 5 | 1683 | |||
BC126314.1 | - | 4, 5 | 1679 |
