siRNA Details
Gene Symbol: LY86-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026970.1 | 5 | 567 | |||
GenBank mRNA | AK091027.1 | - | 5 | 688 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG703873.1 | - | 5 | 491 | Chr. 6 - 6366489 - 6622744 [-] on Build GRCh38 |
