siRNA Details
Gene Symbol: LINC00174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026873.1 | 5 | 1003 | |||
GenBank mRNA | AK124597.1 | - | 1 | 124 | ||
BC020399.1 | - | 5 | 1003 | |||
DB067709.1 | - | 1 | 447 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026873.1 | 5 | 1003 | |||
GenBank mRNA | AK124597.1 | - | 1 | 124 | ||
BC020399.1 | - | 5 | 1003 | |||
DB067709.1 | - | 1 | 447 |