siRNA Details
Gene Symbol: LOC441601 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003034.2 | 1, 2 | 290 | |||
GenBank mRNA | BC067889.1 | - | 1, 2 | 280 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI341338.1 | - | 20 | Chr. 11 - 50280052 - 50295338 [-] on Build GRCh38 |
