siRNA Details
Gene Symbol: INHBA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027118.1 | 1 | 55 | |||
NR_027119.1 | 1 | 23 | ||||
GenBank mRNA | AK096266.1 | - | 37 | |||
BC037931.1 | - | 1 | 60 | |||
BC047446.1 | - | 1 | 22 |
