siRNA Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278302.1 | NP_001265231.1 | 2 | 685 | ||
NM_001278304.1 | NP_001265233.1 | 1 | 1183 | |||
NM_138930.3 | NP_620308.1 | 2 | 685 | |||
GenBank mRNA | AK001399.1 | - | 1 | 1187 | ||
BC095468.1 | - | 2 | 169 | 30557745 | ||
BM746689.1 | - | 2 | 156 | |||
CF128595.1 | - | 2 | 169 |
