siRNA Details
Gene Symbol: MIR1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_031658.1 | 1 | 30 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF744754.1 | - | 3 | 379 | Chr. 20 - 61953469 - 61998178 [-] on Build GRCh38 |
