siRNA Details
Gene Symbol: FAM86B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024361.1 | 1 | 55 | |||
NR_024362.1 | 1 | 55 | ||||
NR_024363.1 | 1 | 55 | ||||
GenBank mRNA | AK307256.1 | - | 1 | 25 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA207233.1 | - | 1 | 55 | Chr. 8 - 8228570 - 8237180 [+] on Build GRCh38 |
