siRNA Details
Gene Symbol: FAM86B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024361.1 | 5 | 403 | |||
NR_024362.1 | 5 | 500 | ||||
NR_024363.1 | 4 | 398 | ||||
GenBank mRNA | AK304567.1 | - | 5 | 472 | ||
AK307256.1 | - | 4 | 368 | |||
BC035792.1 | - | 5 | 378 |
