siRNA Details
Gene Symbol: LINC01144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024270.1 | 1 | 1587 | |||
GenBank mRNA | BC006119.1 | - | 2 | 1078 | ||
BC035497.2 | - | 1 | 1587 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK128109.1 | - | 2 | 3000 | Chr. 19 - 19741548 - 19745256 [-] on Build GRCh38 |
