siRNA Details
Gene Symbol: LINC00310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027266.1 | 1 | 83 | |||
NR_027267.1 | 1 | 83 | ||||
GenBank mRNA | AF426258.1 | - | 1 | 83 | ||
AK092704.1 | - | 1 | 12 | |||
BC093749.1 | - | 1 | 13 | |||
BC111996.1 | - | 1 | 13 |
