siRNA Details
Gene Symbol: BREA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015445.1 | 2 | 964 | |||
GenBank mRNA | AL833948.1 | - | 2 | 964 | ||
AY940073.1 | - | 2 | 1933 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BREA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015445.1 | 2 | 964 | |||
GenBank mRNA | AL833948.1 | - | 2 | 964 | ||
AY940073.1 | - | 2 | 1933 |