siRNA Details
Gene Symbol: LINC01591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026706.2 | 3 | 450 | |||
NR_120395.1 | 3 | 450 | ||||
NR_120396.1 | 2 | 433 | ||||
GenBank mRNA | BC045817.1 | - | 3 | 450 | ||
BC117350.1 | - | 3 | 343 | |||
BC117352.1 | - | 3 | 344 | |||
BC144006.1 | - | 3 | 344 | |||
BC171736.1 | - | 2 | 327 |
