siRNA Details
Gene Symbol: ZNF252P-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026974.1 | 1 | 2553 | |||
GenBank mRNA | AK092777.1 | - | 1 | 2378 | ||
BC046439.1 | - | 1 | 2553 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ185413.1 | - | 1 | 160 | Chr. 8 - 145005204 - 145005938 [+] on Build GRCh38 |
