siRNA Details
Gene Symbol: CECR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024482.1 | 2 | 225 | |||
NR_024483.1 | 2 | 511 | ||||
GenBank mRNA | AF307448.1 | - | 2 | 505 | ||
BI962745.1 | - | 2 | 225 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CECR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024482.1 | 2 | 225 | |||
NR_024483.1 | 2 | 511 | ||||
GenBank mRNA | AF307448.1 | - | 2 | 505 | ||
BI962745.1 | - | 2 | 225 |