siRNA Details
Gene Symbol: LOC440910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030728.1 | 1 | 74 | |||
GenBank mRNA | AK091798.1 | - | 1 | 74 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA584196.1 | - | 1 | 74 | Chr. 2 - 131279290 - 131296893 [+] on Build GRCh38 |
