siRNA Details
Gene Symbol: LOC728613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003713.1 | 3, 4 | 301 | |||
GenBank mRNA | AK311210.1 | - | 3, 4 | 299 | ||
BC014604.1 | - | 3, 4 | 483 | |||
BC045555.1 | - | 3, 4 | 290 | |||
BC053586.1 | - | 6, 7 | 1357 |
