siRNA Details
Gene Symbol: LOC220729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003266.2 | 8 | 969 | |||
GenBank mRNA | AK311713.1 | - | 6 | 674 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC220729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003266.2 | 8 | 969 | |||
GenBank mRNA | AK311713.1 | - | 6 | 674 |