siRNA Details
Gene Symbol: LOC220729 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003266.2 | 8 | 1108 | |||
GenBank mRNA | AK311713.1 | - | 6 | 813 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF551791.1 | - | 5 | 806 | Chr. 3 - 197614382 - 197622320 [-] on Build GRCh38 |
