siRNA Details
Gene Symbol: LINC00847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027183.2 | 3 | 1378 | |||
NR_045678.1 | 3 | 1355 | ||||
NR_045679.1 | 3 | 1360 | ||||
GenBank mRNA | AK095170.1 | - | 2 | 1976 | ||
BC047586.1 | - | 3 | 1292 |
