siRNA Details
Gene Symbol: LINC00964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027321.1 | 4 | 3511 | |||
GenBank mRNA | AK056899.1 | - | 4 | 2135 | ||
BC043249.2 | - | 4 | 3464 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB516601.1 | - | 3 | 370 | Chr. 8 - 124945694 - 124951095 [+] on Build GRCh38 |
