siRNA Details
Gene Symbol: MIR600HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026677.1 | 1 | 2316 | |||
GenBank mRNA | AK123844.1 | - | 1 | 46 | ||
BC015678.2 | - | 1 | 306 | |||
BX648949.1 | - | 1 | 1957 | |||
CR749230.1 | - | 1 | 2315 |
