siRNA Details
Gene Symbol: MIR600HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026677.1 | 1 | 1603 | |||
GenBank mRNA | BX648949.1 | - | 1 | 1244 | ||
CR749230.1 | - | 1 | 1602 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIR600HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026677.1 | 1 | 1603 | |||
GenBank mRNA | BX648949.1 | - | 1 | 1244 | ||
CR749230.1 | - | 1 | 1602 |