siRNA Details
Gene Symbol: MIR600HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026677.1 | 1 | 4093 | |||
GenBank mRNA | AK025814.1 | - | 1 | 1255 | ||
AK123844.1 | - | 1 | 1823 | |||
BC015678.2 | - | 1 | 2083 | |||
BC034918.1 | - | 1 | 45 | |||
BX648949.1 | - | 1 | 3740 | |||
CR749230.1 | - | 1 | 4098 |
