siRNA Details
Gene Symbol: CYCSP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001560.1 | 1 | 190 | |||
GenBank mRNA | R91214.1 | - | 1 | 272 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CYCSP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001560.1 | 1 | 190 | |||
GenBank mRNA | R91214.1 | - | 1 | 272 |