siRNA Details
Gene Symbol: LOC107133515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015341.2 | 6, 7 | 659 | |||
GenBank mRNA | BG721507.1 | - | 6, 7 | 619 | ||
DR004497.1 | - | 5, 6 | 518 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC107133515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015341.2 | 6, 7 | 659 | |||
GenBank mRNA | BG721507.1 | - | 6, 7 | 619 | ||
DR004497.1 | - | 5, 6 | 518 |