siRNA Details
Gene Symbol: PRAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282275.1 | NP_001269204.1 | 2 | 560 | ||
NM_001282276.1 | NP_001269205.1 | 2 | 494 | |||
XM_006721865.3 | XP_006721928.1 | 2 | 1176 | |||
XM_011524747.2 | XP_011523049.1 | 2 | 486 | |||
XM_011524748.1 | XP_011523050.1 | 2 | 431 | |||
GenBank mRNA | AF532777.1 | - | 2 | 548 | ||
BX106051.1 | - | 2 | 494 |
