siRNA Details
Gene Symbol: LINC00910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027412.1 | 4 | 1186 | |||
NR_027413.2 | 4 | 1186 | ||||
GenBank mRNA | AK123585.1 | - | 6 | 587 | ||
AK311685.1 | - | 3 | 483 | |||
AL832479.1 | - | 4 | 600 | |||
BC035366.1 | - | 4 | 833 |
