siRNA Details
Gene Symbol: LINC00910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027412.1 | 1 | 447 | |||
NR_027413.2 | 1 | 447 | ||||
GenBank mRNA | BC035366.1 | - | 1 | 94 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI821852.1 | - | 1 | 93 | Chr. 17 - 43379137 - 43388545 [-] on Build GRCh38 |
