siRNA Details
Gene Symbol: LINC00910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027412.1 | 1, 2 | 606 | |||
NR_027413.2 | 1, 2 | 606 | ||||
GenBank mRNA | AK311685.1 | - | 1, 2 | 43 | ||
AL832479.1 | - | 1, 2 | 20 | |||
BC035366.1 | - | 1, 2 | 253 |
