siRNA Details
Gene Symbol: LINC00910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027412.1 | 3 | 1080 | |||
NR_027413.2 | 3 | 1080 | ||||
GenBank mRNA | AK123585.1 | - | 5 | 481 | ||
AL832479.1 | - | 3 | 494 | |||
BC035366.1 | - | 3 | 727 |
