siRNA Details
Gene Symbol: LINC00910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027412.1 | 2 | 955 | |||
NR_027413.2 | 2 | 955 | ||||
GenBank mRNA | AK123585.1 | - | 4 | 356 | ||
AK311685.1 | - | 2 | 392 | |||
AL832479.1 | - | 2 | 369 | |||
BC035366.1 | - | 2 | 602 |
