siRNA Details
Gene Symbol: TTC41P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027249.1 | 13 | 3461 | |||
GenBank mRNA | BC064342.1 | - | 3 | 351 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG114468.1 | - | 348 | Chr. - - [-] on Build |
