siRNA Details
Gene Symbol: LINC00598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024505.1 | 5 | 614 | |||
NR_024506.1 | 5 | 614 | ||||
NR_024507.2 | 5 | 614 | ||||
GenBank mRNA | AY116214.1 | - | 5 | 614 | ||
AY116215.1 | - | 1 | 21 | |||
AY116216.1 | - | 1 | 21 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI520193.1 | - | 4 | 443 | Chr. 13 - 40450931 - 40481006 [-] on Build GRCh38 |
